Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-chloroaniline 95%

Supplier: AMBEED, INC
Warning

A102371-1G A102371-5G A102371-100G A102371-100MG A102371-10G A102371-250MG A102371-25G A102371-1G A102371-5G
103796-362EA 8 USD InStock
103796-362 103822-056 76809-306 76809-308 76809-310 76809-312 76809-314 77308-940 77308-942
3-Bromo-2-chloroaniline 95%
3-Bromo-2-chloroaniline
Precaución: for Research Use Only


Formula: C₆H₅BrClN
MW: 206.468 g/mol
Boiling Pt: 267.1±20.0 °C (760 mm Hg)
Melting Pt: 31…33 °C
Flash Pt: 115.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11848536
CAS Number: 56131-46-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty