Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-ethylbenzoic acid 97%

Supplier: AMBEED, INC
Warning

A275641-1G A275641-5G A275641-100MG A275641-10G A275641-5G A275641-1G
103813-272EA 99 USD InStock
103813-272 103853-196 77014-510 77014-512 77093-052 77305-208
3-Bromo-2-ethylbenzoic acid 97%
3-Bromo-2-ethylbenzoic acid
Precaución: for Research Use Only


Formula: C₉H₉BrO₂
Storage Temperature: Ambient
MDL Number: MFCD17676588
CAS Number: 99548-72-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty