Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-ethylpyridine 98%

Supplier: AMBEED, INC
Warning

A332819-1G A332819-5G A332819-1G A332819-5G
103825-618EA 62 USD InStock
103825-618 103836-582 77306-440 77306-442
3-Bromo-2-ethylpyridine 98%
3-Bromo-2-ethylpyridine
Precaución: for Research Use Only


Formula: C₇H₈BrN
Storage Temperature: Ambient
MDL Number: MFCD13189503
CAS Number: 38749-81-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty