Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-fluorobenzonitrile 98%

Supplier: AMBEED, INC
Warning

A110914-1G A110914-5G A110914-100G A110914-10G A110914-250MG A110914-25G A110914-1G A110914-5G A110914-500G
103797-750EA 7 USD InStock
103797-750 103823-558 76687-564 76687-566 76687-568 76687-570 77310-356 77310-358 77876-828
3-Bromo-2-fluorobenzonitrile 98%
3-Bromo-2-fluorobenzonitrile
Precaución: for Research Use Only


Formula: C₇H₃BrFN
MW: 200.01
Boiling Pt: 221.8 °C (760 mm Hg)
Melting Pt: 37…40 °C
Flash Pt: 87.983 °C
Storage Temperature: Ambient
MDL Number: MFCD06657980
CAS Number: 840481-82-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty