Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-methoxybenzonitrile 98%

Supplier: AMBEED, INC
Warning

A345429-1G A345429-25G A345429-5G A345429-100G A345429-10G A345429-250MG A345429-1G
103825-924EA 23 USD InStock
103825-924 103836-906 76839-966 76839-968 76876-222 76876-224 76876-226 77306-566
3-Bromo-2-methoxybenzonitrile 98%
3-Bromo-2-methoxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₆BrNO
Melting Pt: 75…78 °C
Storage Temperature: Ambient
MDL Number: MFCD09834773
CAS Number: 874472-98-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty