Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2-methyl-6-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A190175-1G A190175-5G A190175-250MG A190175-1G
103816-270EA 69 USD InStock
103816-270 103846-870 77097-122 77302-216
3-Bromo-2-methyl-6-nitropyridine 97%
3-Bromo-2-methyl-6-nitropyridine
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
Boiling Pt: 299.5±35.0 °C (760 mm Hg)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD28130027
CAS Number: 1231930-13-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty