Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-bromo-2-oxobutanoate 90%

Supplier: AMBEED, INC
Warning

A118302-1G A118302-100MG A118302-250MG A118302-1G A118302-5G
103799-032EA 74 USD InStock
103799-032 103799-300 76971-348 76971-350 77311-710 77311-712
Ethyl-3-bromo-2-oxobutanoate 90%
Ethyl-3-bromo-2-oxobutanoate
Precaución: for Research Use Only


Formula: C₆H₉BrO₃
Boiling Pt: 213 °C (760 mm Hg)
Flash Pt: 83 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19440735
CAS Number: 57332-84-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty