Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-(bromomethyl)benzonitrile 96%

Supplier: AMBEED, INC
Danger

A754884-5G A754884-10G A754884-250MG A754884-1G A754884-5G
103840-868EA 218 USD InStock
103840-868 76820-332 76820-334 77308-262 77308-264
3-Bromo-4-(bromomethyl)benzonitrile 96%
3-Bromo-4-(bromomethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₅Br₂N
MW: 274.9400 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09261034
CAS Number: 89892-39-7
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty