Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-4-chloropyridine 98%

Supplier: AMBEED, INC
Danger

A320850-1G A320850-25G A320850-5G A320850-10G A320850-500G A320850-1G A320850-100G
103825-252EA 5 USD InStock
103825-252 103836-184 76839-066 76839-068 77290-286 77290-288 77306-226 77748-627
3-Bromo-4-chloropyridine 98%
3-Bromo-4-chloropyridine
Precaución: for Research Use Only


Formula: C₅H₃BrClN
Boiling Pt: 207.3 °C (760 mm Hg)
Flash Pt: 79.2 °C
Storage Temperature: Freezer
MDL Number: MFCD00234008
CAS Number: 36953-42-1
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty