Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-chloropyrazolo[1,5-a]pyrimidine 98%

Supplier: AMBEED, INC
Warning

A373429-5G A373429-10G A373429-250MG A373429-1G A373429-25G A373429-5G
103826-534EA 15 USD InStock
103826-534 103837-552 76788-714 76788-716 76840-818 76840-820 76840-822
3-Bromo-5-chloropyrazolo[1,5-a]pyrimidine 98%
3-Bromo-5-chloropyrazolo[1,5-a]pyrimidine
Precaución: for Research Use Only


Formula: C6H3BrClN3
MW: 232.4700 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11520857
CAS Number: 960613-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty