Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-methylaniline 98%

Supplier: AMBEED, INC
Warning

A259368-1G A259368-5G A259368-100G A259368-250MG A259368-1G A259368-25G A259368-5G
103812-438EA 6 USD InStock
103812-438 103852-318 76786-944 76786-946 77091-874 77091-876 77091-878
3-Bromo-5-methylaniline 98%
3-Bromo-5-methylaniline
Precaución: for Research Use Only


Formula: C7H8BrN
MW: 186.05 g/mol
Boiling Pt: 257.5 °C (760 mm Hg)
Flash Pt: 111.7 °C
Storage Temperature: Ambient
MDL Number: MFCD11845553
CAS Number: 74586-53-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty