Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-methylpyridin-2-ol 97%

Supplier: AMBEED, INC
Warning

A197426-5G A197426-1G A197426-5G
103816-920EA 9 USD InStock
103816-920 103847-542 77085-552 77085-554
3-Bromo-5-methylpyridin-2-ol 97%
3-Bromo-5-methylpyridin-2-ol
Precaución: for Research Use Only


Formula: C6H6BrNO
MW: 188.02 g/mol
Boiling Pt: 324 °C (760 mm Hg)
Melting Pt: 78…82 °C
Storage Temperature: Ambient
MDL Number: MFCD03791312
CAS Number: 17282-02-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty