Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-5-nitrophenol 95%

Supplier: AMBEED, INC
Danger

A247766-1G A247766-5G A247766-250MG A247766-1G
103811-794EA 14 USD InStock
103811-794 103851-630 76815-582 77090-970
3-Bromo-5-nitrophenol 95%
3-Bromo-5-nitrophenol
Precaución: for Research Use Only


Formula: C₆H₄BrNO₃
Boiling Pt: 310.4 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09965960
CAS Number: 116632-23-6
UN: 3077
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty