Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromocatechol 97%

Supplier: AMBEED, INC
Warning

A194773-1G A194773-5G A194773-10G A194773-1G A194773-25G A194773-5G A194773-100G
103816-692EA 6 USD InStock
103816-692 103847-304 76785-072 77085-228 77085-230 77085-232 77593-252
3-Bromocatechol 97%
3-Bromocatechol
Precaución: for Research Use Only


Formula: C₆H₅BrO₂
Boiling Pt: 242.6 °C (760 mm Hg)
Melting Pt: 40…41 °C
Flash Pt: 101 °C
Storage Temperature: Ambient
MDL Number: MFCD00869768
CAS Number: 14381-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty