Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromobenzamidine hydrochloride 97%

Supplier: AMBEED, INC
Warning

A152674-1G A152674-5G A152674-250MG A152674-5G A152674-1G
103804-416EA 19 USD InStock
103804-416 103842-588 77137-836 77299-966 77318-214
3-Bromobenzamidine hydrochloride 97%
3-Bromobenzamidine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₈BrClN₂
MW: 235.51
Boiling Pt: 307.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114439
CAS Number: 16796-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty