Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromobenzoic acid 98%

Supplier: AMBEED, INC
Warning

A216787-5G A216787-100G A216787-10G A216787-500G A216787-25G
103849-304EA 5 USD InStock
103849-304 76814-774 76814-776 76814-778 77087-908
3-Bromobenzoic acid 98%
3-Bromobenzoic acid
Precaución: for Research Use Only


Formula: C₇H₅BrO₂
MW: 201.02 g/mol
Boiling Pt: 308
Melting Pt: 155
Flash Pt: 140.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00002487
CAS Number: 585-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty