Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-bromofuran-2,5-dione 97%

Supplier: AMBEED, INC
Warning

A413119-100G A413119-10G A413119-250MG A413119-25G A413119-1G A413119-5G
103827-256EA 14 USD InStock
103827-256 103838-302 76694-876 76694-878 76694-880 76694-882 76841-854 76841-856
3-bromofuran-2,5-dione 97%
3-bromofuran-2,5-dione
Precaución: for Research Use Only


Formula: C₄HBrO₃
MW: 176.953 g/mol
Boiling Pt: 215 °C (760 mm Hg)
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00005519
CAS Number: 5926-51-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty