Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromoisoxazole-5-carboxylic acid 97%

Supplier: AMBEED, INC
Warning

A520946-1G A520946-5G A520946-250MG A520946-1G
103828-504EA 50 USD InStock
103828-504 103839-612 76843-614 77279-772 77307-714
3-Bromoisoxazole-5-carboxylic acid 97%
3-Bromoisoxazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C4H2BrNO3
Boiling Pt: 375.1 °C (760 mm Hg)
Flash Pt: 180.692 °C
Storage Temperature: Freezer
MDL Number: MFCD07368611
CAS Number: 6567-35-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty