Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromophthalic acid 97%

Supplier: AMBEED, INC
Warning

A432741-1G A432741-5G A432741-250MG A432741-10G
103827-536EA 14 USD InStock
103827-536 103838-602 76842-300 76842-302 77033-944 77872-691
3-Bromophthalic acid 97%
3-Bromophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅BrO₄
Boiling Pt: 408.8 °C (760 mm Hg)
Melting Pt: 177…178 °C
Flash Pt: 201.1 °C
Storage Temperature: Ambient
MDL Number: MFCD17069368
CAS Number: 116-69-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty