Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Bromo-2,6-diaminopyridine 95%

Supplier: AMBEED, INC
Warning

A197563-1G A197563-5G A197563-10G A197563-250MG
103816-940EA 24 USD InStock
103816-940 103847-560 77030-034 77030-036
3-Bromo-2,6-diaminopyridine 95%
3-Bromo-2,6-diaminopyridine
Precaución: for Research Use Only


Formula: C₅H₆BrN₃
Boiling Pt: 294.2±35.0 °C (760 mm Hg)
Flash Pt: 131.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04113819
CAS Number: 54903-86-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty