Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-fluoropyridine 98%

Supplier: AMBEED, INC
Warning

A295609-1G A295609-5G A295609-25G A295609-100G A295609-10G A295609-1G
103814-066EA 5 USD InStock
103814-066 103835-200 76837-860 77154-112 77154-114 77305-676
3-Chloro-2-fluoropyridine 98%
3-Chloro-2-fluoropyridine
Precaución: for Research Use Only


Formula: C₅H₃ClFN
MW: 131.54
Boiling Pt: 162.2 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD04114144
CAS Number: 1480-64-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty