Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-methoxy-5-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A245247-1G A245247-5G A245247-1G A245247-5G
103811-630EA 26 USD InStock
103811-630 103851-468 76987-748 76987-750 77090-712 77090-714
3-Chloro-2-methoxy-5-nitropyridine 97%
3-Chloro-2-methoxy-5-nitropyridine
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₃
MW: 188.569 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18254096
CAS Number: 22353-53-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty