Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A266065-100G A266065-10G A266065-500G A266065-25G
103852-678EA 7 USD InStock
103852-678 77276-676 77276-678 77276-680 77304-944
3-Chloro-o-toluic acid 98%
3-Chloro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇ClO₂
MW: 170.6
Boiling Pt: 290.9 °C (760 mm Hg)
Melting Pt: 163…166 °C
Flash Pt: 129.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00053312
CAS Number: 7499-08-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty