Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-2-nitroaniline 98%

Supplier: AMBEED, INC
Warning

A112559-1G A112559-5G A112559-100G A112559-10G A112559-25G A112559-500G A112559-1G A112559-5G
103798-036EA 10 USD InStock
103798-036 103823-874 77094-062 77094-064 77094-066 77094-068 77310-678 77310-680
3-Chloro-2-nitroaniline 98%
3-Chloro-2-nitroaniline
Precaución: for Research Use Only


Formula: C₆H₅ClN₂O₂
MW: 172.57
Boiling Pt: 327.1 °C (760 mm Hg)
Melting Pt: 107…108 °C
Flash Pt: 151.6 °C
Storage Temperature: Ambient
MDL Number: MFCD01317803
CAS Number: 59483-54-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty