Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4,5-dihydroxybenzoic acid 95%

Supplier: AMBEED, INC
Warning

A297049-1G A297049-5G A297049-1G A297049-100MG
103814-124EA 391 USD InStock
103814-124 103835-266 76837-952 77014-884
3-Chloro-4,5-dihydroxybenzoic acid 95%
3-Chloro-4,5-dihydroxybenzoic acid
Precaución: for Research Use Only


Formula: C₇H₅ClO₄
Boiling Pt: 391.7 °C (760 mm Hg)
Flash Pt: 190.7 °C
Storage Temperature: Ambient
MDL Number: MFCD01742816
CAS Number: 87932-49-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty