Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4-nitropyridine 97%

Supplier: AMBEED, INC
Warning

A255960-1G A255960-5G A255960-5G A255960-250MG A255960-1G
103812-226EA 31 USD InStock
103812-226 103852-086 77091-556 77276-430 77304-660
3-Chloro-4-nitropyridine 97%
3-Chloro-4-nitropyridine
Precaución: for Research Use Only


Formula: C5H3ClN2O2
MW: 158.54 g/mol
Boiling Pt: 256 °C (760 mm Hg)
Melting Pt: 25…26 °C
Flash Pt: 108 °C
Storage Temperature: Freezer
MDL Number: MFCD00490146
CAS Number: 13194-60-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty