Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-fluorobenzene -1,2-diamine 98%

Supplier: AMBEED, INC
Warning

A325659-1G A325659-250MG A325659-5G
103825-412EA 52 USD InStock
103825-412 103836-358 76828-270 76839-304
3-Chloro-5-fluorobenzene -1,2-diamine 98%
3-Chloro-5-fluorobenzene -1,2-diamine
Precaución: for Research Use Only


Formula: C₆H₆ClFN₂
Storage Temperature: Refrigerator
MDL Number: MFCD20723094
CAS Number: 153505-33-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty