Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-5-methoxyaniline 98%

Supplier: AMBEED, INC
Warning

A126041-1G A126041-250MG A126041-1G A126041-25G A126041-5G
103800-756EA 46 USD InStock
103800-756 76870-482 77312-924 77312-926 77312-928
3-Chloro-5-methoxyaniline 98%
3-Chloro-5-methoxyaniline
Precaución: for Research Use Only


Formula: C₇H₈ClNO
MW: 157.598 g/mol
Boiling Pt: 280.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD11035922
CAS Number: 10272-06-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty