Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-6,11-dihydro-6-methyl-5,5,11-trioxodibenzo[c,f][1,2]thiazepine 95%

Supplier: AMBEED, INC
Warning

A160073-1G A160073-5G A160073-1G A160073-5G
103805-328EA 9 USD InStock
103805-328 103843-558 77319-472 77319-474
3-Chloro-6,11-dihydro-6-methyl-5,5,11-trioxodibenzo[c,f][1,2]thiazepine 95%
3-Chloro-6,11-dihydro-6-methyl-5,5,11-trioxodibenzo[c,f][1,2]thiazepine
Precaución: for Research Use Only


Formula: C14H10ClNO3S
Boiling Pt: 494.6 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD07368146
CAS Number: 26638-53-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty