Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chlorobenzaldehyde 97%

Supplier: AMBEED, INC
Warning

A103016-100G A103016-25G A103016-500G
103807-852EA 120 USD InStock
103807-852 77107-314 77107-316 77107-318
3-Chlorobenzaldehyde 97%
3-Chlorobenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅ClO
MW: 140.57 g/mol
Boiling Pt: 214
Melting Pt: 9…12 °C
Flash Pt: 88 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003350
CAS Number: 587-04-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty