Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-N,N-diethylpropan-1-amine 95%

Supplier: AMBEED, INC
Danger

A373070-1G A373070-25G A373070-5G A373070-10G A373070-1G
103826-524EA 7 USD InStock
103826-524 103837-542 76840-802 76840-804 77278-448 77306-848
3-Chloro-N,N-diethylpropan-1-amine 95%
3-Chloro-N,N-diethylpropan-1-amine
Precaución: for Research Use Only


Formula: C₇H₁₆ClN
Boiling Pt: 169…171 °C (760 mm Hg)
Flash Pt: 56.9 °C
Storage Temperature: Freezer
MDL Number: MFCD00039385
CAS Number: 104-77-8
UN: 2734
ADR: 8,3,Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty