Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyano-1H-pyrazole-5-carboxylic acid 95%

Supplier: AMBEED, INC
Danger

A411525-1G A411525-1G A411525-250MG
103827-228EA 371 USD InStock
103827-228 77307-150 77673-002
3-Cyano-1H-pyrazole-5-carboxylic acid 95%
3-Cyano-1H-pyrazole-5-carboxylic acid
Precaución: for Research Use Only


Formula: C₅H₃N₃O₂
Boiling Pt: 489.3±30.0 °C (760 mm Hg)
Flash Pt: 249.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD09036046
CAS Number: 1187361-13-2
UN: 3439
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty