Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyanobenzenesulphonyl chloride 98%

Supplier: AMBEED, INC
Danger

A538490-1G A538490-5G A538490-250MG A538490-10G
103828-612EA 12 USD InStock
103828-612 103839-734 76843-774 76843-776 76980-146 77882-308
3-Cyanobenzenesulphonyl chloride 98%
3-Cyanobenzenesulphonyl chloride
Precaución: For research use only


Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Boiling Pt: 148 °C (3 mm Hg)
Melting Pt: 49…53 °C
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8 Ⅱ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty