Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Cyanopropanoic acid 95%

Supplier: AMBEED, INC
Warning

A149484-1G A149484-5G A149484-250MG A149484-1G A149484-5G
103804-046EA 18 USD InStock
103804-046 103842-164 77272-934 77317-664 77317-666
3-Cyanopropanoic acid 95%
3-Cyanopropanoic acid
Precaución: for Research Use Only


Formula: C4H5NO2
MW: 99.0880 g/mol
Boiling Pt: 313.5 °C (760 mm Hg)
Melting Pt: 43…47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00233974
CAS Number: 16051-87-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty