Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Decylthiophene 98%

Supplier: AMBEED, INC
Warning

A147934-1G A147934-5G A147934-25G A147934-1G A147934-5G A147934-10G A147934-100G
103803-850EA 5 USD InStock
103803-850 103841-966 77299-616 77317-384 77317-386 77871-724 77871-723
3-Decylthiophene 98%
3-Decylthiophene
Precaución: for Research Use Only


Formula: C₁₄H₂₄S
Boiling Pt: 299.4 °C (760 mm Hg)
Flash Pt: 99.1 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00143180
CAS Number: 65016-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty