Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid 98%

Supplier: AMBEED, INC
Warning

A150850-1G A150850-5G A150850-100MG A150850-250MG A150850-1G A150850-5G A150850-10G
103804-206EA 52 USD InStock
103804-206 103842-346 77095-806 77095-808 77317-878 77317-880 77685-256
3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid 98%
3-Ethoxy-2,2-dimethyl-3-oxopropanoic acid
Precaución: for Research Use Only


Formula: C7H12O4
Boiling Pt: 250.5 °C (760 mm Hg)
Flash Pt: 97.7 °C
Storage Temperature: Ambient
MDL Number: MFCD04003823
CAS Number: 5471-77-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty