Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Ethynylpiperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A131328-1G A131328-100MG A131328-250MG A131328-1G
103801-568EA 91 USD InStock
103801-568 77027-072 77027-074 77314-066
3-Ethynylpiperidine hydrochloride 95%
3-Ethynylpiperidine hydrochloride
Precaución: for Research Use Only


Formula: C₇H₁₂ClN
Storage Temperature: Refrigerator
MDL Number: MFCD12546352
CAS Number: 959918-19-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty