Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-1H-indazole 95%

Supplier: AMBEED, INC
Warning

A113033-1G A113033-100MG A113033-250MG
103798-148EA 232 USD InStock
103798-148 103823-984 76982-826 76982-828
3-Fluoro-1H-indazole 95%
3-Fluoro-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₅FN₂
Storage Temperature: Refrigerator
MDL Number: MFCD09909613
CAS Number: 66607-26-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty