Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-2-(trifluoromethyl)aniline 98%

Supplier: AMBEED, INC
Warning

A653287-100MG A653287-10G A653287-250MG A653287-1G A653287-25G A653287-5G
103840-408EA 56 USD InStock
103840-408 76775-568 76775-570 76775-572 76844-582 76844-584 76844-586
3-Fluoro-2-(trifluoromethyl)aniline 98%
3-Fluoro-2-(trifluoromethyl)aniline
Precaución: for Research Use Only


Formula: C₇H₅F₄N
Boiling Pt: 199.2 °C (760 mm Hg)
Flash Pt: 89 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06658260
CAS Number: 123973-22-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty