Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-o-toluic acid 98%

Supplier: AMBEED, INC
Warning

A194163-1G A194163-5G A194163-25G A194163-5G A194163-10G A194163-1G A194163-100G
103816-626EA 5 USD InStock
103816-626 103847-236 77085-156 77085-158 77236-992 77302-394 77593-246
3-Fluoro-o-toluic acid 98%
3-Fluoro-o-toluic acid
Precaución: for Research Use Only


Formula: C₈H₇FO₂
Boiling Pt: 261.2 °C (760 mm Hg)
Melting Pt: 158…160 °C
Flash Pt: 111.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00075246
CAS Number: 699-90-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty