Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(hydroxymethyl)phenol 95%

Supplier: AMBEED, INC
Warning

A142638-1G A142638-1G A142638-250MG
103803-200EA 65 USD InStock
103803-200 77299-306 77871-690
3-Fluoro-4-(hydroxymethyl)phenol 95%
3-Fluoro-4-(hydroxymethyl)phenol
Precaución: for Research Use Only


Formula: C₇H₇FO₂
MW: 142.13 g/mol
Boiling Pt: 291.0±25.0 °C (760 mm Hg)
Flash Pt: 129.8 °C
Storage Temperature: Ambient
MDL Number: MFCD06797932
CAS Number: 96740-92-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty