Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(trifluoromethoxy)aniline 97%

Supplier: AMBEED, INC
Danger

A120580-1G A120580-100G A120580-100MG A120580-250MG A120580-25G A120580-1G A120580-5G A120580-10G
103799-958EA 10 USD InStock
103799-958 76781-722 76781-724 76781-726 76781-728 77312-064 77312-066 77871-526
3-Fluoro-4-(trifluoromethoxy)aniline 97%
3-Fluoro-4-(trifluoromethoxy)aniline
Precaución: for Research Use Only


Formula: C₇H₅F₄NO
Storage Temperature: Refrigerator
MDL Number: MFCD09832383
CAS Number: 1017779-69-9
UN: 2810
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty