Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-formylphenylboronic acid 98%

Supplier: AMBEED, INC
Warning

A338620-5G A338620-1G A338620-5G A338620-100G A338620-10G A338620-250MG
103825-772EA 11 USD InStock
103825-772 103836-744 76839-768 76839-770 77142-056 77142-058 77142-060
3-Fluoro-4-formylphenylboronic acid 98%
3-Fluoro-4-formylphenylboronic acid
Precaución: for Research Use Only


Formula: C₇H₆BFO₃
MW: 167.93 g/mol
Boiling Pt: 349.3 °C (760 mm Hg)
Melting Pt: 249…250 °C
Flash Pt: 165.1 °C
Storage Temperature: Ambient
MDL Number: MFCD02093051
CAS Number: 248270-25-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty