Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-p-anisidine 97%

Supplier: AMBEED, INC
Warning

A180123-5G A180123-100G A180123-25G A180123-5G
103845-864EA 10 USD InStock
103845-864 76769-540 77083-344 77083-346
3-Fluoro-p-anisidine 97%
3-Fluoro-p-anisidine
Precaución: for Research Use Only


Formula: C₇H₈FNO
MW: 141.15 g/mol
Boiling Pt: 253.4 °C (760 mm Hg)
Melting Pt: 81…83 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00075040
CAS Number: 366-99-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty