Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-(trifluoromethyl)benzonitrile 97%

Supplier: AMBEED, INC
Danger

A278102-1G A278102-5G A278102-100G A278102-10G A278102-25G A278102-5G A278102-1G
103813-390EA 6 USD InStock
103813-390 103853-328 76772-264 76772-266 77093-238 77093-240 77305-288
3-Fluoro-4-(trifluoromethyl)benzonitrile 97%
3-Fluoro-4-(trifluoromethyl)benzonitrile
Precaución: for Research Use Only


Formula: C₈H₃F₄N
MW: 189.11 g/mol
Boiling Pt: 212
Melting Pt: 45…48 °C
Flash Pt: 96 °C
Storage Temperature: Ambient
MDL Number: MFCD00236280
CAS Number: 231953-38-1
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty