Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-hydroxybenzaldehyde 95%

Supplier: AMBEED, INC
Warning

A258885-1G A258885-5G A258885-250MG A258885-1G A258885-5G
103812-406EA 35 USD InStock
103812-406 103852-284 76874-800 77091-814 77091-816
3-Fluoro-5-hydroxybenzaldehyde 95%
3-Fluoro-5-hydroxybenzaldehyde
Precaución: for Research Use Only


Formula: C₇H₅FO₂
Boiling Pt: 246.2±20.0 °C (760 mm Hg)
Flash Pt: 102.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185579
CAS Number: 1023290-12-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty