Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-5-methylpyridin-2-amine 95%

Supplier: AMBEED, INC
Warning

A101312-1G A101312-5G A101312-250MG A101312-1G
103795-904EA 129 USD InStock
103795-904 103821-590 76970-150 77297-054
3-Fluoro-5-methylpyridin-2-amine 95%
3-Fluoro-5-methylpyridin-2-amine
Precaución: for Research Use Only


Formula: C₆H₇FN₂
MW: 126.134 g/mol
Boiling Pt: 195.5±35.0 °C (760 mm Hg)
Flash Pt: 72 °C
Storage Temperature: Refrigerator
MDL Number: MFCD13185333
CAS Number: 1211590-31-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty