Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluorophthalic acid 97%

Supplier: AMBEED, INC
Warning

A258985-1G A258985-5G A258985-100G A258985-10G A258985-25G A258985-1G
103812-414EA 6 USD InStock
103812-414 103852-292 76771-828 76771-830 77091-828 77304-758
3-Fluorophthalic acid 97%
3-Fluorophthalic acid
Precaución: for Research Use Only


Formula: C₈H₅FO₄
MW: 184.12 g/mol
Boiling Pt: 367.9 °C (760 mm Hg)
Melting Pt: 175…177 °C
Flash Pt: 181.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00039524
CAS Number: 1583-67-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty