Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Formyl-4-hydroxybenzonitrile 95%

Supplier: AMBEED, INC
Warning

A136752-1G A136752-100MG A136752-250MG A136752-5G
103802-346EA 113 USD InStock
103802-346 77272-292 77272-294 77298-866
3-Formyl-4-hydroxybenzonitrile 95%
3-Formyl-4-hydroxybenzonitrile
Precaución: for Research Use Only


Formula: C₈H₅NO₂
Boiling Pt: 241.3 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09839541
CAS Number: 74901-29-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty