Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Hydrazinylpyridine hydrochloride 98%

Supplier: AMBEED, INC
Warning

A542252-1G A542252-100G A542252-10G A542252-5G
103828-642EA 7 USD InStock
103828-642 103839-762 76818-990 76818-992 76843-814
3-Hydrazinylpyridine hydrochloride 98%
3-Hydrazinylpyridine hydrochloride
Precaución: for Research Use Only


Formula: C₅H₈ClN₃
Flash Pt: 141.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06797881
CAS Number: 650638-17-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty